Product Details

SNP ID
rs1951392
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:103387030 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTCAATCTAGTTGTTGACTATAT[G/T]CTGTAAAACATCTAGTAAATAATTT
Phenotype
MIM: 602678
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MARK3 PubMed Links

Gene Details

Gene
MARK3
Gene Name
microtubule affinity regulating kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128918.2 Intron NP_001122390.2
NM_001128919.2 Intron NP_001122391.2
NM_001128920.2 Intron NP_001122392.2
NM_001128921.2 Intron NP_001122393.2
NM_002376.6 Intron NP_002367.5
XM_005267641.4 Intron XP_005267698.2
XM_005267642.4 Intron XP_005267699.2
XM_005267643.4 Intron XP_005267700.2
XM_006720146.3 Intron XP_006720209.1
XM_011536769.2 Intron XP_011535071.1
XM_017021291.1 Intron XP_016876780.1
XM_017021292.1 Intron XP_016876781.1
XM_017021293.1 Intron XP_016876782.1
XM_017021294.1 Intron XP_016876783.1
XM_017021295.1 Intron XP_016876784.1
XM_017021296.1 Intron XP_016876785.1
XM_017021297.1 Intron XP_016876786.1
XM_017021298.1 Intron XP_016876787.1
XM_017021299.1 Intron XP_016876788.1
XM_017021300.1 Intron XP_016876789.1
XM_017021301.1 Intron XP_016876790.1
XM_017021302.1 Intron XP_016876791.1
XM_017021303.1 Intron XP_016876792.1
XM_017021304.1 Intron XP_016876793.1
XM_017021305.1 Intron XP_016876794.1
XM_017021306.1 Intron XP_016876795.1
XM_017021307.1 Intron XP_016876796.1
XM_017021308.1 Intron XP_016876797.1
XM_017021309.1 Intron XP_016876798.1
XM_017021310.1 Intron XP_016876799.1
XM_017021311.1 Intron XP_016876800.1

View Full Product Details