Product Details

SNP ID
rs7225550
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:48147881 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTTACATTTCAGGGAACTTGCTAA[C/T]CTGGAGATTGATGGGCCTGGATACA
Phenotype
MIM: 604969
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR1203 PubMed Links

Gene Details

Gene
MIR1203
Gene Name
microRNA 1203
There are no transcripts associated with this gene.

Gene
SKAP1
Gene Name
src kinase associated phosphoprotein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001075099.1 Intron NP_001068567.1
NM_003726.3 Intron NP_003717.3
XM_005257755.3 Intron XP_005257812.1
XM_017025257.1 Intron XP_016880746.1
XM_017025258.1 Intron XP_016880747.1
XM_017025259.1 Intron XP_016880748.1
XM_017025260.1 Intron XP_016880749.1

View Full Product Details