Product Details

SNP ID
rs2232816
Assay Type
Validated
NCBI dbSNP Submissions
87
Location
Chr.1:171783899 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
CCAGATGAGCTTCTTGAAGATGGAC[A/G]TGACGCAGATGGAGTTTCCTGATGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
METTL13 PubMed Links

Gene Details

Gene
METTL13
Gene Name
methyltransferase like 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007239.1 327 Missense Mutation ATG,GTG M105V NP_001007240.1
NM_014955.2 327 Missense Mutation ATG,GTG M19V NP_055770.1
NM_015935.4 327 Missense Mutation ATG,GTG M105V NP_057019.3
XM_017001455.1 327 Missense Mutation ATG,GTG M19V XP_016856944.1
XM_017001456.1 327 Missense Mutation ATG,GTG M19V XP_016856945.1
XM_017001457.1 327 Missense Mutation ATG,GTG M19V XP_016856946.1
XM_017001458.1 327 Missense Mutation ATG,GTG M105V XP_016856947.1
XM_017001459.1 327 Missense Mutation ATG,GTG M19V XP_016856948.1

View Full Product Details