Product Details

SNP ID
rs2292507
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:52544580 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GTGCACTCAGGCTGGAACCCTTCCC[C/T]AGGGTGTCTTTGTAATCGTTGGCAC
Phenotype
MIM: 608245
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRT71 PubMed Links

Gene Details

Gene
KRT71
Gene Name
keratin 71
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033448.2 1594 Silent Mutation CTA,CTG L508L NP_258259.1
XM_017018749.1 1594 Silent Mutation CTA,CTG L426L XP_016874238.1

View Full Product Details