Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001271634.1 | Intron | NP_001258563.1 | |||
NM_001271635.1 | Intron | NP_001258564.1 | |||
NM_005444.2 | Intron | NP_005435.1 | |||
XM_011512138.2 | Intron | XP_011510440.1 | |||
XM_017005248.1 | Intron | XP_016860737.1 | |||
XM_017005249.1 | Intron | XP_016860738.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_020935.2 | Intron | NP_065986.2 | |||
XM_005246720.2 | Intron | XP_005246777.1 | |||
XM_005246721.3 | Intron | XP_005246778.1 | |||
XM_005246722.3 | Intron | XP_005246779.1 | |||
XM_005246724.1 | Intron | XP_005246781.1 | |||
XM_011511538.2 | Intron | XP_011509840.1 | |||
XM_011511541.2 | Intron | XP_011509843.1 | |||
XM_017004577.1 | Intron | XP_016860066.1 |