Product Details

SNP ID
rs1046715
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:86941181 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAAACCTTTCCCTATGTTTTATC[C/T]CCTGAAAAAAAAAATGTATGGAATA
Phenotype
MIM: 609980
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VGLL3 PubMed Links

Gene Details

Gene
VGLL3
Gene Name
vestigial like family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320493.1 7045 Intron NP_001307422.1
NM_001320494.1 7045 UTR 3 NP_001307423.1
NM_016206.3 7045 UTR 3 NP_057290.2
XM_006713138.3 7045 UTR 3 XP_006713201.1

View Full Product Details