Product Details

SNP ID
rs11882386
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35937806 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGACCCCTGCAACCCTTACAGCCT[C/T]TCGCTGTCTATCCGGTCCGACCGTC
Phenotype
MIM: 612809
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105372383 PubMed Links

Gene Details

Gene
LOC105372383
Gene Name
uncharacterized LOC105372383
There are no transcripts associated with this gene.

Gene
LRFN3
Gene Name
leucine rich repeat and fibronectin type III domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024509.1 Intron NP_078785.1
XM_017027302.1 Intron XP_016882791.1

View Full Product Details