Product Details

SNP ID
rs613316
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65427784 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTTTTCAGAACTCCTTGTTTCTC[A/G]TCCTGTTTTTTATGATTACAAAACT
Phenotype
MIM: 612769
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
NEAT1 PubMed Links

Gene Details

Gene
NEAT1
Gene Name
nuclear paraspeckle assembly transcript 1 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details