Product Details

SNP ID
rs595108
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62795763 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTGGAGAGGTAGGCCAAGCCAA[C/G]ACGGCTTGGGGGCAGAATGGGAGAA
Phenotype
MIM: 602647 MIM: 602946
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MIR6514 PubMed Links

Gene Details

Gene
MIR6514
Gene Name
microRNA 6514
There are no transcripts associated with this gene.

Gene
MIR6748
Gene Name
microRNA 6748
There are no transcripts associated with this gene.

Gene
NXF1
Gene Name
nuclear RNA export factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081491.1 Intron NP_001074960.1
NM_006362.4 Intron NP_006353.2
Gene
TAF6L
Gene Name
TATA-box binding protein associated factor 6 like
There are no transcripts associated with this gene.

Gene
TMEM179B
Gene Name
transmembrane protein 179B
There are no transcripts associated with this gene.

Gene
TMEM223
Gene Name
transmembrane protein 223
There are no transcripts associated with this gene.

View Full Product Details