Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
smgl-1
Entrez Gene ID
172561
Gene Name
Uncharacterized protein
Gene Aliases
CELE_F20G4.1
Chromosome Location
Chr.I: 7918438 - 7926503 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383416.1 7 - 8 1858 94

View Full Product Details