Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
col-54
Entrez Gene ID
185226
Gene Name
Col_cuticle_N domain-containing protein
Gene Aliases
CELE_F33D11.3
Chromosome Location
Chr.I: 5842709 - 5844062 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059293.4 NP_491694.1 1 - 2 150 144

View Full Product Details