Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- png-1
- Entrez Gene ID
-
173028
- Gene Name
- Peptide-N(4)-(N-acetyl-beta-glucosaminyl)asparagine amidase
- Gene Aliases
-
CELE_F56G4.5
- Chromosome Location
-
Chr.I: 11380231 - 11388751 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_060512.5 |
NP_492913.1 |
6 - 7 |
1240 |
63 |
View Full Product Details