Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fkh-6
Entrez Gene ID
181907
Gene Name
Fork-head domain-containing protein
Gene Aliases
CELE_B0286.5
Chromosome Location
Chr.II: 4379585 - 4383406 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062374.3 NP_494775.1 1 - 2 155 63

View Full Product Details