Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
col-76
Entrez Gene ID
174320
Gene Name
Col_cuticle_N domain-containing protein
Gene Aliases
CELE_M110.1
Chromosome Location
Chr.II: 8204341 - 8207495 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063325.6 NP_495726.2 4 - 5 1080 70

View Full Product Details