Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-13A8
Entrez Gene ID
188358
Gene Name
Putative cytochrome P450 CYP13A8
Gene Aliases
CELE_T10B9.4
Chromosome Location
Chr.II: 9805600 - 9807569 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063714.2 NP_496115.1 5 - 6 1197 170

View Full Product Details