Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pxl-1
Entrez Gene ID
175831
Gene Name
protein-coding
Gene Aliases
CELE_C28H8.6
Chromosome Location
Chr.III: 5889721 - 5896017 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026014.4 NP_001021185.2 9 - 10 966 79
NM_001129205.3 NP_001122677.1 7 - 8 789 79
GenBank mRNA EU239658.1 - 9 - 10 961 79
EU239660.1 - 7 - 8 778 79

View Full Product Details