Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
set-22
Entrez Gene ID
179457
Gene Name
SET domain-containing protein
Gene Aliases
CELE_Y32F6A.1
Chromosome Location
Chr.V: 10437003 - 10441159 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380744.2 NP_001366641.1 6 - 7 690 126

View Full Product Details