Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srw-17
Entrez Gene ID
188395
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_T10H4.6
Chromosome Location
Chr.V: 15275440 - 15277015 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074382.2 NP_506783.1 2 - 3 1059 130

View Full Product Details