Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fkh-9
Entrez Gene ID
180670
Gene Name
protein-coding
Gene Aliases
CELE_K03C7.2
Chromosome Location
Chr.X: 3816847 - 3830113 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029589.4 NP_001024760.1 1 - 2 113 82
NM_001380939.1 NP_001367812.1 2 - 3 234 82

View Full Product Details