Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ift-81
Entrez Gene ID
185183
Gene Name
IFT81_CH domain-containing protein
Gene Aliases
CELE_F32A6.2
Chromosome Location
Chr.X: 5298561 - 5306542 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076499.9 NP_508900.6 5 - 6 496 95

View Full Product Details