Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hbl-1
Entrez Gene ID
180848
Gene Name
protein-coding
Gene Aliases
CELE_F13D11.2
Chromosome Location
Chr.X: 5822142 - 5827755 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047778.5 NP_001041243.1 3 - 4 447 65
NM_001047779.5 NP_001041244.1 2 - 3 246 65
GenBank mRNA AF097737.1 - 3 - 4 438 65

View Full Product Details