Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
coh-1
Entrez Gene ID
181003
Gene Name
protein-coding
Gene Aliases
CELE_K08A8.3
Chromosome Location
Chr.X: 7465193 - 7475019 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322622.4 NP_001309700.1 3 - 4 323 82
NM_001322623.4 NP_001309701.1 2 - 3 242 82
NM_076861.7 NP_509262.2 8 - 9 1630 82

View Full Product Details