Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-17
Entrez Gene ID
191719
Gene Name
Nuclear Hormone Receptor family
Gene Aliases
CELE_C02B4.2
Chromosome Location
Chr.X: 12663760 - 12667854 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077714.5 NP_510115.1 2 - 3 182 106

View Full Product Details