Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD86
Entrez Gene ID
403764
Gene Name
CD86 molecule
Gene Aliases
-
Chromosome Location
Chr.33: 25256267 - 25318855 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003146.2 NP_001003146.1 4 - 5 717 89
XM_014110137.2 XP_013965612.1 4 - 5 869 89
GenBank mRNA AF106826.1 - 4 - 5 705 89

View Full Product Details