Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UQCC2
Entrez Gene ID
100683221
Gene Name
ubiquinol-cytochrome c reductase complex assembly factor 2
Gene Aliases
C12H6orf125, MNF1
Chromosome Location
Chr.12: 3050329 - 3063831 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003431698.4 XP_003431746.1 2 - 3 263 71

View Full Product Details