Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KCNS2
Entrez Gene ID
481979
Gene Name
potassium voltage-gated channel modifier subfamily S member 2
Gene Aliases
-
Chromosome Location
Chr.13: 622676 - 628301 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005627816.3 XP_005627873.1 - 1505 151

View Full Product Details