Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pex19
Entrez Gene ID
100689302
Gene Name
peroxisomal biogenesis factor 19
Gene Aliases
Peroxin-19, Pxf
Chromosome Location
Chr.Un|NW_003613783: 638890 - 645380 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001246736.1 NP_001233665.1 5 - 6 599 67
GenBank mRNA JI878699.1 - 5 - 6 614 67
JP046069.1 - 5 - 6 614 67
U05959.1 - 5 - 6 599 67

View Full Product Details