Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mbd3l1
Entrez Gene ID
100770293
Gene Name
methyl-CpG binding domain protein 3-like 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615026: 38676 - 39250 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003511084.2 XP_003511132.1 1 - 1 421 94

View Full Product Details