Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nacad
Entrez Gene ID
100769774
Gene Name
NAC alpha domain containing
Gene Aliases
I79_011878
Chromosome Location
Chr.Un|NW_003614085: 1052348 - 1061100 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007645281.1 XP_007643471.1 4 - 5 4096 58

View Full Product Details