Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdr2l
Entrez Gene ID
100758231
Gene Name
cerebellar degeneration-related protein 2-like
Gene Aliases
I79_000945
Chromosome Location
Chr.Un|NW_003613598: 491830 - 507425 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007643388.1 XP_007641578.1 2 - 3 165 80

View Full Product Details