Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mc5r
Entrez Gene ID
100758289
Gene Name
melanocortin 5 receptor
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614248: 353926 - 354900 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007647013.1 XP_007645203.1 1 - 1 722 72

View Full Product Details