Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mbl2
Entrez Gene ID
100769668
Gene Name
mannose-binding lectin (protein C) 2, soluble
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613782: 2051412 - 2054810 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003500228.1 XP_003500276.1 3 - 4 372 65

View Full Product Details