Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Efcab4a
Entrez Gene ID
100754254
Gene Name
EF-hand calcium binding domain 4A
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614763: 311575 - 316290 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509769.2 XP_003509817.1 7 - 8 2019 67

View Full Product Details