Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ikbip
Entrez Gene ID
100757532
Gene Name
IKBKB interacting protein
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614719: 40083 - 54877 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509493.2 XP_003509541.1 2 - 3 364 89
GenBank mRNA JP056565.1 - 1 - 2 97 89

View Full Product Details