Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gsc
Entrez Gene ID
100760478
Gene Name
goosecoid homeobox
Gene Aliases
I79_007754
Chromosome Location
Chr.Un|NW_003613846: 206725 - 211211 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007642039.1 XP_007640229.1 2 - 3 240 95

View Full Product Details