Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
METTL7B
Entrez Gene ID
102179920
Gene Name
methyltransferase like 7B
Gene Aliases
-
Chromosome Location
Chr.5: 56935390 - 56937888 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005680399.3 XP_005680456.1 1 - 2 802 61
GenBank mRNA JO424572.1 - 1 - 2 4993 61

View Full Product Details