Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rps27a
Entrez Gene ID
100286787
Gene Name
ribosomal protein S27a
Gene Aliases
Ubiz
Chromosome Location
Chr.Un|NT_176371: 9379563 - 9381715 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001173066.1 NP_001166537.1 4 - 5 342 69
XM_005002637.3 XP_005002694.1 5 - 6 394 69
GenBank mRNA D83209.1 - 4 - 5 342 69

View Full Product Details