Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bsh
Entrez Gene ID
35266
Gene Name
brain-specific homeobox
Gene Aliases
BSH, Bsh, CG10604, Dmel\CG10604, Dmel_CG10604, E86, Hox11-310, lambdaE86
Chromosome Location
Chr.2L: 19765776 - 19769067 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0000529
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058002.3 NP_477350.2 3 - 4 1159 76
NM_206006.2 NP_995728.2 3 - 4 1159 76
GenBank mRNA BT022203.2 - 3 - 4 1144 76

View Full Product Details