Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Ir25a
- Entrez Gene ID
-
33683
- Gene Name
- Ionotropic receptor 25a
- Gene Aliases
-
CG15627, CT35778, DmelIR25a, Dmel\CG15627, Dmel_CG15627, IR25a, ir25a
- Chromosome Location
-
Chr.2L: 4830846 - 4835300 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0031634
- Gene Symbol
- tank
- Entrez Gene ID
-
33684
- Gene Name
- CG15626-PA
- Gene Aliases
-
CG15626, Dmel\CG15626, Dmel_CG15626
- Chromosome Location
-
Chr.2L: 4835335 - 4837999 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0031635
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BT099717.1 |
- |
8 - 9 |
2707 |
85 |
View Full Product Details