Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tw
Entrez Gene ID
31024
Gene Name
twisted
Gene Aliases
CG12311, DPOMT2, DmPOMT2, Dmel\CG12311, Dmel_CG12311, EG:34F3.7, POMT-2, POMT2, Pomt2, Tw, dPOMT2, dPomt2
Chromosome Location
Chr.X: 784746 - 788215 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0086368
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272173.1 NP_001259102.1 8 - 9 1821 93
NM_130502.3 NP_569858.1 8 - 9 1821 93
GenBank mRNA AB176551.1 - 8 - 9 1662 93
AY095525.1 - 8 - 9 1776 93

View Full Product Details