Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- okr
- Entrez Gene ID
-
33507
- Gene Name
- okra
- Gene Aliases
-
CG3736, DhR54, DmRAD54, DmRad54, Dmel\CG3736, Dmel_CG3736, Dmrad54, OKR, Okr, RAD54, RAD54L, Rad54, Ras54, rad54
- Chromosome Location
-
Chr.2L: 2989594 - 2992759 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0002989
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057313.5 |
NP_476661.1 |
2 - 3 |
1367 |
91 |
View Full Product Details