Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- dpr5
- Entrez Gene ID
-
41381
- Gene Name
- defective proboscis extension response 5
- Gene Aliases
-
CG5308, CT16895, Dmel\CG5308, Dmel_CG5308, Dpr-5, Dpr5
- Chromosome Location
-
Chr.3R: 11499984 - 11505951 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0037908
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_141823.3 |
NP_650080.3 |
1 - 2 |
437 |
81 |
GenBank mRNA |
AY047569.1 |
- |
1 - 2 |
437 |
81 |
View Full Product Details