Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cht10
Entrez Gene ID
3355116
Gene Name
Chitinase 10
Gene Aliases
BcDNA:LP04696, CG17682, CG18140, CG18140-PA3, CHT1, CHT3, Cht1, Cht3, DmCHT1, DmCHT10, DmCHT3, DmCht1, DmCht10, DmCht3, Dmel\CG18140, Dmel_CG18140
Chromosome Location
Chr.2L: 22999574 - 23025915 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0250907
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042957.2 NP_001036422.1 5 - 6 3870 117
NM_001273768.1 NP_001260697.1 5 - 6 3954 117
NM_001273769.1 NP_001260698.1 5 - 6 3941 117
GenBank mRNA BT021372.1 - 5 - 6 3870 117

View Full Product Details