Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dlx6a
Entrez Gene ID
30586
Gene Name
distal-less homeobox 6a
Gene Aliases
dlx6
Chromosome Location
Chr.19: 41465094 - 41467029 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131323.1 NP_571398.1 1 - 2 504 127
GenBank mRNA BC114264.1 - 1 - 2 526 127
U67844.1 - 1 - 2 504 127

View Full Product Details