Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ywhabb
- Entrez Gene ID
-
406419
- Gene Name
- tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, beta polypeptide b
- Gene Aliases
-
wu:fb22g04, wu:fr86a12, ywhab, ywhab2, zgc:64092
- Chromosome Location
-
Chr.8: 45418985 - 45430801 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_213145.1 |
NP_998310.1 |
2 - 3 |
361 |
146 |
GenBank mRNA |
BC053247.1 |
- |
2 - 3 |
361 |
146 |
View Full Product Details