Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rmnd5b
Entrez Gene ID
393747
Gene Name
required for meiotic nuclear division 5 homolog B
Gene Aliases
zgc:73277
Chromosome Location
Chr.14: 49220100 - 49244543 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200774.1 NP_957068.1 7 - 8 910 106
GenBank mRNA AY394935.1 - 6 - 7 899 106
BC059606.1 - 7 - 8 910 106

View Full Product Details