Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dacha
Entrez Gene ID
192121
Gene Name
dachshund a
Gene Aliases
cb907
Chromosome Location
Chr.21: 34363972 - 34658298 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152955.2 NP_694487.2 9 - 10 1716 86
XM_005161406.4 XP_005161463.1 9 - 10 1743 86
XM_009295592.3 XP_009293867.1 9 - 10 1752 86
XM_009295593.3 XP_009293868.1 9 - 10 1752 86
XM_021468748.1 XP_021324423.1 9 - 10 1752 86
GenBank mRNA AF427108.1 - 9 - 10 1717 86
BC162471.1 - 9 - 10 1629 86

View Full Product Details