Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cracr2b
Entrez Gene ID
790936
Gene Name
calcium release activated channel regulator 2B
Gene Aliases
efcab4a, zgc:158448
Chromosome Location
Chr.25: 3830845 - 3868033 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080081.1 NP_001073550.1 9 - 10 1099 96
GenBank mRNA BC128877.1 - 9 - 10 1099 96

View Full Product Details