Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bms1
Entrez Gene ID
553241
Gene Name
BMS1 ribosome biogenesis factor
Gene Aliases
bms1l, im:7152071, wu:fb37d08
Chromosome Location
Chr.12: 315985 - 346901 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111150.2 NP_001104620.2 10 - 10 1562 158
GenBank mRNA BC078263.1 - 10 - 10 1561 164
BC134022.1 - 10 - 10 1563 164
BC155256.1 - 10 - 10 1549 164
BC155771.1 - 11 - 11 76

View Full Product Details