Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LYZ
Entrez Gene ID
396218
Gene Name
lysozyme (renal amyloidosis)
Gene Aliases
LYZC
Chromosome Location
Chr.1: 35542896 - 35546583 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205281.1 NP_990612.1 3 - 4 409 86
GenBank mRNA CR390743.1 - 3 - 4 86
V00428.1 - - 409 86

View Full Product Details